Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Contrad 70, liquid detergent, Decon Labs
Supplier: Decon Labs
Contrad
1003 1003 1005 1004 1004 1002 1005 1002
89233-154EA 764.66 USD InStock
CA89233-156 CA42000-000 CA89233-154 89233-154 CA89233-152 42000-000 89233-156 89233-152
Contrad 70, liquid detergent, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
Contrad 70 liquid detergent concentrate is phosphate-free, chlorine-free, and biodegradable
Cleans glass, plastic, ceramic, and ferrous metals by soaking, manual scrubbing, or ultrasonic cleaning. Contrad 70 is free-rinsing and disposable straight to the drain. Made with recycled packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty