Would you like to log in again?
Contrad 100, Low-Foaming Liquid Detergent, Decon Labs
Supplier: Decon Labs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Contrad
1506 1506 1504 1505 1503 1504 1503
89233-160EA 1221.11 USD InStock
89233-160 CA89233-158 89233-158 89166-410 89166-412 CA89166-412 CA89233-160
Contrad 100, Low-Foaming Liquid Detergent, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
Contrad 100 is a low-foaming alkaline liquid detergent formulated for Clean-In-Place systems
Effective in removing a wide variety of contaminants from stainless steel, glass, and plastic surfaces. Its low-foaming characteristics prevent pump cavitation and excessive residual foam. Contrad 100 is free-rinsing, phosphate-free, and disposable straight to the drain. Made with recycled packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty