Would you like to log in again?
Bel-Art Magic Touch 2™ Ice Buckets, ATS Life Sciences
Supplier: Bel-Art Products, a part of SP
Bel-Art
M16807-2002 M16807-2003 M16807-2004 M16807-2001 M16807-4004 M16807-4003 M16807-4002 M16807-4001
89233-346EA 156.33 USD InStock
89233-346 89233-348 89233-342 89233-352 89233-344 89233-354 89233-340 89233-350
Bel-Art Magic Touch 2™ Ice Buckets, ATS Life Sciences
Ice Containers
Superior insulating properties combined with high chemical resistance make Magic Touch 2™ Ice Buckets compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries, or liquid nitrogen.
  • Excellent insulation
  • High chemical resistance
  • Convenient pour spout
  • Stackable
  • Lightweight and durable
  • Expanded urethane construction
  • Temperature range: –196...+100 °C (–320... +212 °F)
Able to withstand a wide temperature range, these high-quality, expanded urethane, laboratory ice buckets are lightweight, durable, non-sweating and impervious to moisture and odors.
Ordering information: Magic Touch 2™ Ice Buckets are available in 2.5L and 4.0L sizes, each in four colors; ebony, cherry red, true blue and emerald green.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty