Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Disposable Sterile Animal Feeding/Oral Gavage Needles, Cadence Science®
Supplier: Cadence Science
9929 9924B 9931 9924 9916 9918 9927B 9927 9922B 9925 9917 9923B 9926B 9932 9930 9915 9928 9933 9923 9922 9926 9928B 9925B 9921 9920
89233-514PK 1047.04 USD InStock
89233-514 89233-516 89233-510 89233-512 89233-496 89233-498 89233-492 20068-666 20068-668 89233-494 89233-490 89233-508 89233-504 89233-506 89233-500 89233-522 89233-488 89233-524 89233-502 20068-670 89233-520 89233-486 89233-480 89233-482 89233-518
Disposable Sterile Animal Feeding/Oral Gavage Needles, Cadence Science®
Animal Supplies Animal Feeding Needles
Disposable sterile feeding needles are for critical applications
Available in three styles: malleable stainless steel, flexible plastic (PTFE), and flexible plastic (PTFE) with stainless steel bite protectors. All needles feature a plastic luer hub, and a soft silicone tip. They are malleable to any angle. Non-toxic and non-pyrogenic.
Ordering information: Packaged in sterile peel pouches, 5 needles per pouch, 100 needles per box.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty