Would you like to log in again?
GLOVE SURG PI 9.0 PF PK50PR
Supplier: Cardinal Health
Made with Synthetic Polyisoprene. Sterile, Powder-free Surgical Gloves^ Sterilized by gamma irradiation. Available in hand-specific sizes 9. Independent thumb for greater movement and less hand strain. Beaded cuff and length of 30.5cm (12Inch). Average thickness: 10mil.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE SURG PI 9.0 PF PK50PR 89233-844 2D72PL90X Case of 400 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty