Would you like to log in again?
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FlashGel®
57034 57031 50473 57068 57025 57061 57023 57064 50462 57022 57060 57067 57033 57051 57069 57069 57034 57050 57063 57029 57028 57031 57064 57026 57027 57062 57025 57022 57029 50473 57063 57065 57068 50462 57051 50577 57031 57060 57023 57033 50577 57032 57050 57024 57040 57024 57026 57067 57028 57062 50475 57027 57065 57032 50475 57040
89233-898EA 879.04 USD InStock
CA95053-312 CA95015-612 CA95053-310 CA95015-614 89233-898 CA95053-314 CA95015-616 CA95015-618 89400-690 89400-692 89400-694 CA95045-602 95015-634 95053-314 95053-312 95015-632 CA89233-898 95015-638 CA95045-604 95015-636 95053-310 89135-718 95015-640 CA95015-634 95045-604 95015-624 95015-623 CA95015-636 95015-622 95045-602 CA95015-630 95015-628 CA95015-632 95015-626 CA95015-638 CA89135-718 CA95015-640 CA89400-688 95015-630 CA89400-686 95015-612 CA95015-624 CA95015-626 CA95015-620 95015-616 CA95015-622 95015-614 95015-618 CA95015-628 CA89400-692 CA89400-690 CA11011-026 CA89400-694 89400-686 95015-620 89400-688
FlashGel™ DNA, RNA and Recovery Systems
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
FlashGel Systems consist of enclosed, disposable, precast agarose gel cassettes and a combination electrophoresis and transilluminator unit.
  • Systems provide nearly instant results
  • Simply load samples, watch bands migrate and get data in as little as two minutes
FlashGel DNA system separates DNA in 2–7 minutes with outstanding resolution and allows monitoring of gel runs right at the bench without UV light
FlashGel RNA system is a fast and sensitive tool for analysis of sample integrity and optimized for the unique requirements of RNA. Recommended for verification and analysis of total RNA, quick checks of native RNA, and checking RNA degradation and mRNA purity. The system provides results in 30 minutes or less while avoiding use of hazardous reagents and contaminating RNases. RNA fragments are analyzed on the FlashGel® system using the same fast and simple procedure used for DNA. Separation is complete in less than eight minutes and RNA samples are ready for imaging within 10 to 20 minutes. FlashGel® system for RNA is 5 to 20 times more sensitive than ethidium bromide gels and RNA <10 ng per band are clearly detected, conserving precious RNA samples.
FlashGel system for DNA recovery eliminates agarose gel preparation and delivers highly efficient recovery, free from inhibitors and UV-induced damage, in a simple 5 to 10 minute protocol. Go from sample loading to recovery in just five minutes. Recover samples directly from the cassette without band excision or purification. Visualize sample recovery without UV light and recover at 80 to 100% efficiency.
FlashGel Dock is an electrophoresis apparatus with a built-in transilluminator that provides visualization of both DNA and RNA cassettes. The FlashGel camera captures gel images from the FlashGel system right at the benchtop with complete gel run and image capture in just 5 minutes. Camera software is compatible with PC computers.
The highly sensitive system also allows 5 to 20 times the reduction in DNA levels, saving both time and money.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty