Would you like to log in again?
SP Bel-Art Test Tube Capper, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F18799-0000
89233-990EA 53.06 USD InStock
89233-990
SP Bel-Art Test Tube Capper, Bel-Art Products, a part of SP
Crimpers, Cappers and Decappers
The vinyl-covered handhold fits comfortably in the palm of the hand while the polypropylene tip presses down on the cap to securely position it within the test tube.
  • Vinyl-covered handhold
  • Polypropylene tip secures cap in position
  • Reduced finger soreness and hand stress
Capping can be performed on glass or plastic test tubes in a rack, or while being held in the hand
With the Scienceware® Test Tube Capper, you can use your entire hand for leverage, reducing finger soreness and hand stress.
It is designed for use with recessed center test tube caps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty