Would you like to log in again?
SP Bel-Art Compact Support Stand, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
H18303-0005
89233-992EA 99.13 USD InStock
89233-992
SP Bel-Art Compact Support Stand, Bel-Art Products, a part of SP
Support Stands
This non-corrosive support stand with stainless steel mounting rod provides modern advantages over metal ring stands.
  • Compact base design
  • Polypropylene construction with stainless steel mounting rod
  • Excellent stability
  • Non-corrosive
  • Easy to clean
  • Autoclavable
The compact polypropylene base has a smooth surface making cleaning easy, and measures 21 x 16 cm (8¼ x 6¼”) x 4.5 cmH (1¾”).
At a weight of 1.95kg (4.3 lb), it provides excellent stability and can be used to support a variety of types of plasticware and labware. The 9.4 mm (3/8”) diameter x 50 cm (19¾”) long stainless steel rod can be mounted at the center or at one end of the base. Autoclavable.
Ordering information: The funnel, ring clamp, and flask for this unit are sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty