Would you like to log in again?
HisTrap™ Excel Affinity Chromatography Columns
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Histrap™
17371205 29-0485-86 17371206 17371205 17-3712-06 29-0485-86 17371206 17-3712-05 17371205
89501-372EA 87.45 USD InStock
89501-372 CA89233-414 89233-412 CA89233-412 89233-414 CA89234-482 89234-480 CA89234-480 CA11033-012
HisTrap™ Excel Affinity Chromatography Columns
Chromatography Columns
HisTrap excel columns are prepacked with Ni Sepharose excel affinity resins for capture and purification of histidine-tagged proteins secreted into eukaryotic cell culture supernatants by immobilized metal ion affinity chromatography (IMAC).
  • Load eukaryotic cell culture samples containing secreted histidine-tagged proteins directly with retained binding capacity
  • Increase target protein yield and decrease degradation through reduced and simplified sample handling
  • HisTrap excel columns allow direct purification of cell-free, unclarified samples
HisTrap excel 1 ml and 5 ml are ready-to-use IMAC columns prepacked with Ni Sepharose excel. The design of the columns in combination with the specific properties of the resin enables fast and convenient purifications.
The special type of filter in the top and bottom of the columns makes it possible to load large volumes of cell-free, unclarified samples directly on the columns without causing back pressure problems. This time-saving property helps prevent degradation and loss of sensitive target proteins.
HisTrap excel columns are made of biocompatible polypropylene.
HisTrap excel columns can be operated either with a peristaltic pump or with chromatography systems such as ÄKTA systems. The columns cannot be opened or refilled.
Delivery information: Supplied with a stopper on the inlet and a snap-off end on the outlet.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty