Would you like to log in again?
VWR® Disposable Sterile Face Shield
Supplier: VWR
89234-522
89234-522CS 1621.45 USD InStock
89234-522
VWR® Disposable Sterile Face Shield
Face Shields
Disposable ETO sterile faceshields are made of transparent anti-fog polyethylene film with an elastic band.
  • Transparent anti-fog polyethylene construction
  • Elastic band
  • Provides long-lasting comfort
Face shields are individually packaged, and provide long-lasting comfort with a secure fit.
Application: laboratory, electric, or pharmaceutical cleanroom.
Certifications: ISO9001, ISO13485, ISO14001. CE marked. GMP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty