Would you like to log in again?
Contrex® AP, Powder Detergent, Decon Labs
Supplier: Decon Labs
Contrex®
5225 5204 5225 5204
89234-820EA 279.97 USD InStock
CA89234-820 89234-820 89234-818 CA89234-818
Contrex® AP, Powder Detergent, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
Contrex® AP is a concentrated powdered labware detergent formulated for use in manual, soak or ultrasonic cleaning of labware, scientific apparatus and process equipment
Contrex® AP is completely soluble, free-rinsing and safe to dispose straight to drain. The user friendly bottle allows for easy dispensing and eliminates caking. Also available in a 1/2 ounce single serve which is pre-measured to make a 2 quart solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty