Would you like to log in again?
Contrex® AW, Powder Detergent, Decon Labs
Supplier: Decon Labs
Contrex®
5604 5625 5625 5604
89234-832EA 288.28 USD InStock
CA89234-830 89234-832 CA89234-832 89234-830
Contrex® AW, Powder Detergent, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
Contrex® AW is a concentrated low-foaming powdered cleanser formulated for use in all automatic labware washing machines
Contrex® AW is completely soluble, free-rinsing and biodegradable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty