Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
CiDecon® Plus Wipes, Surface Disinfectant Wipes, Decon Labs
Supplier: Decon Labs
8511
89234-842CS 633.83 USD InStock
89234-842
CiDecon® Plus Wipes, Surface Disinfectant Wipes, Decon Labs
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
CiDecon® Plus Wipes are pre-moistened hard surface disinfectant wipes for use on hard surfaces.
They come in a convenient 'pop-up' canister, are economical, durable and display superior efficacy against a broad range of pathogenic organisms. Achieves hospital disinfection in 10 minutes and maintains continual residual bacteriostatic activity for up to 6 months (see label for details). FDA compliant for use on medical equipment and instruments. CiDecon is also effective against a wide range of organisms including TB. EPA registered.
Certifications: Meets OSHA's Blood Borne Pathogens Standard.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty