Synonyms:
Ethyl alcohol
V1055
V1001GTP
V1016GTP
V1005
V1055TP
V1016G
V1005MTP
V1001TP
V1016TP
V1016
V1055M
V1001
V1005M
V1005HC
V1001G
V1005SGTP
V1005HCTP
V1005TP
V1055MTP
V1005SG
89426-252EA
122.01
USD
InStock
89426-252
89125-176
89125-188
71002-422
71001-866
89125-174
71001-624
89125-186
89125-172
89234-848
89125-192
89125-170
71001-628
89125-190
89234-850
71002-426
89501-404
71001-862
71002-512
89234-852
Ethanol absolute, KOPTEC, meets analytical specification of BP, Ph. Eur., USP (200 Proof)
Ethanol
Pure (undenatured) anhydrous (200 proof/100%) ethyl alcohol meets USP and Multicompendial specifications used as a cleaner, solvent, or reagent.
- Total alcohol: 99.5%
- Ethyl alcohol: 99.5%
- Denaturant: none
Used in research, healthcare, industrial, and production. 200 proof ethanol is commonly referred to as "anhydrous." Koptec's water specification is less than 0.1%.
Ordering information: Tax-paid ethanol prices include the federal excise tax on pure ethanol. To purchase tax-free ethanol, customers must possess an Industrial Alcohol User Permit from TTB specifically marked for the withdrawal and use of alcohol free of tax.