Would you like to log in again?
Socorex Twister™ Universal Pipette Work Station, DWK Life Sciences
Supplier: DWK Life Sciences
W870162 W870158 W870152 W870154 W870156 W870160 W870164
89236-610CS 252.59 USD InStock
89236-610 89236-612 89236-614 89236-616 89236-606 89236-618 89236-608
Socorex Twister™ Universal Pipette Work Station, DWK Life Sciences
Pipette Stands
The stand allows 360° rotation for easy access to the right pipette
It holds 6 pipettes of most brands. The stand is easy to clean and disassemble.
Ordering information: Available in seven translucent colors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty