Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Hazard BinLiner™ Bags, Micronova
Supplier: Micronova
BL2-25REIRBB BL4-4060REPC
89236-646CS 407.09 USD InStock
89236-646 75779-994
Hazard BinLiner™ Bags, Micronova
Bags Waste Disposal Bags
These low density polyethylene Hazard BinLiner™ Bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.
  • Low density polyethylene
  • Hazardous material bags are labeled for waste segregation in the U.S.
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Can be used with BLSSH series BinLiner™ Stands
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas
The Biohazard liners are designed for containment and disposal of biomedical waste and the Hazardous Material liners are designed for segregation and disposal of hazardous waste. Biohazard bags are clearly marked with a Biohazard symbol in black ink.
Certifications: Certificate of Irradiation and Certificate of Conformance shipped with each order. Validation paperwork available upon request. Minimum irradiation dose: 25 kGy.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty