Would you like to log in again?
Sterile Pre-wetted Sealed Edge Cleanroom Laundered 100% Polyester Knit Wiper, Berkshire
Supplier: Berkshire
SSP120.01.10WFI
89237-584CS 1443.76 USD InStock
89237-584
Sterile Pre-wetted Sealed Edge Cleanroom Laundered 100% Polyester Knit Wiper, Berkshire
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Sterile SatPax® MicroSeal® WFI combines MicroSeal® 1200 ultrasonically sealed edge cleanroom laundered wipers composed of 100% continuous filament polyester knit fabric with a high saturation level of 70% IPA and 30% WFI water.
This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents, maintenance of squirt bottles and inconsistent wetting and cleaning associated with wetting a dry wiper. Each package is then gamma irradiated sterile to a 10⁻⁶ Sterility Assurance Level making the product ideal for cleaning critical sterile processing environments.
Pre-wetted with consistent 70% IPA/ 30% WFI (Water for Injection) to a high saturation level. Details: expiration date, radiation dosage, lot information, Meets cGMP requirements for traceability.WFI (Water for Injection) solution for critical applications requiring WFI.Designed for use in wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required.
Certifications: ANSI/AAMI/ISO procedures have been followed for verification of irradiation dose with quarterly audits being performed to monitor bioburden levels and dose. The Sterility Assurance Level (SAL) or the probability of a viable organism being present on a product unit after sterilization is a SAL of 10⁻⁶ (which is the chance of 1 non sterile unit in 1,000,000). - Complies with RoHS and REACH requirements.
Ordering information: 30 shts/pk, 10 pks/cs -Double packed to ensure cleanliness. Quality control and lot traceability identification on each case. Expiration date and Lot Number embossed on each pack. Certificate of Sterility with each case and available on the web by lot number 24/7.
Packaging: Re-sealable solvent resistant packaging.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty