Would you like to log in again?
Medical Gas Storage Cabinets, SECURALL®
Supplier: Securall
MG121FL MG321FL MG104FL MG309H MG309 MG109H MG321 MG306HFL MG102 MG109FL MG104 MG109HFL MG309FL MG102FL MG309HFL MG106HFL MG121 MG304FL MG306H MG109 MG304 MG106H
89237-692EA 3462 USD InStock
89237-692 89237-690 89237-688 89237-710 89237-722 89237-700 89237-686 89237-698 89237-720 89237-684 89237-696 89237-682 89237-694 89237-718 89237-708 89237-716 89237-706 89237-714 89237-704 89237-712 89237-724 89237-702
Medical Gas Storage Cabinets, SECURALL®
Furniture Storage Cabinets
These lockable gas storage cabinets limit access to authorized personnel, protecting cylinders from theft, damage and/or tampering.
Healthcare organizations such as hospitals, nursing homes, and emergency medical services will be prepared for their 3-year JCAHO accreditation survey when their oxygen cylinders are stored safely in SECURALL® MedGas Cabinets. SECURALL® proudly manufactures the highest quality safety storage products available. SECURALL® Gas Cylinder Storage Cabinets are available for storage of D, E, and H cylinders in varying capacities to meet any storage need. The "Capacity_NA" represents Storage capacity. The "Storage" represents Storage types.
The heavy-gauge, double-walled construction shields your cylinders from heat, sparks and flame, while separating them from flammable and combustible materials. Designed for D- and E-size cylinders up to 33-3/4" long x 4-1/4" diameter, and H-size cylinders up to 60” long x 9-1/4” diameter.
Certifications: SMaRT Certified, Meets NFPA 99, 101, 30 & OSHA Standards.
Ordering information: Standard Color is Med Gas Green, please contact your VWR Furniture Specialist for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty