Would you like to log in again?
Weatherproof Flammable Safety Cabinets, SECURALL®
Supplier: Securall
A130WP1 A360WP1 A145WP1 A190WP1 A345WP1 A160WP1 A330WP1 A390WP1
89237-758EA 5312.1 USD InStock
89237-758 89237-748 89237-756 89237-746 89237-754 89237-744 89237-752 89237-750
Weatherproof Flammable Safety Cabinets, SECURALL®
Furniture Storage Cabinets
Double-wall weatherproof cabinets are all-welded, 18-gauge construction using galvannealed steel and painted with a weather-resistant, impact-resistant urethane finish that is also resistant to ultra-violet rays.
All Cabinets STANDARD Features a sloped roof with a 2³/₄" overhang along with interior and exterior astragals to prevent rain penetration. Weatherproof cabinets come standard with corrosion resistant flush-mounted stainless steel locking handle, but may be upgraded to galvanized high security hasp lock. Easily move these cabinets to the appropriate work location using built-in heavy gauge fork lift channels and secure them with fully-welded 7-gauge steel Bolt-Down Brackets with ⁷/₈" diameter opening. Available on all standard cabinet models with manual and self-closing door styles.
Certifications: SMaRT Certified; Meets NFPA 30 and OSHA Regulations. Cabinets meet: EPA 40 CFR 264, 265, NFPA Code 30 and OSHA 1910 requirements.
Ordering information: Standard Color is Yellow, please contact your Avantor Furniture Specialist for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty