Would you like to log in again?
Drum Spill Stations, SECURALL®
Supplier: Securall
DSS12 DSS04 DSS02 DSS06 DSS09
89237-806EA 3139.18 USD InStock
89237-806 89237-804 89237-812 89237-810 89237-808
Drum Spill Stations, SECURALL®
Drum Storage
Controlling unwanted spills is easier than ever with SECURALL® Drum Spill Pallets
Built to provide a lifetime of service, these secondary containment Drum Spill Stations provide an easy and safe solution to organizing hazardous waste in drums. Rugged all-welded 12-gauge galvannealed steel offers long term use. Our Drum Spill Stations are specifically designed to be used as a holding area for drums, dispensing station or waste collection center. All units feature steel planking and forklift channels for easy mobility. Durable enough to be transported at full capacity.
Certifications: SMaRT Certified; Complies with EPA Spill Containment Requirements (40CFR264.175).
Ordering information: Standard Color is Gray, please contact your Avantor Furniture Specialist for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty