Would you like to log in again?
Corning® Gosselin™ Octagonal PET Bottles, Sterile, Corning
Supplier: Corning
Corning®
P1000B-50 P250B-50
89238-026CS 273.05 USD InStock
89238-026 89238-023
Corning® Gosselin™ Octagonal PET Bottles, Sterile, Corning
Bottles Plastic Bottles
These storage bottles are made from break resistant polyethylene terephthalate (PET) with transparency equivalent to glass.
  • Molded graduations
  • Ergonomic shape for easy handling
  • Leak proof, tamper evident HDPE screw cap
Available with Sterility Assurance Level (SAL) of 10⁻³.
With wide mouth opening and designed to collect samples or store laboratory fluids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty