Would you like to log in again?
VWR REFRIGERATOR SWG DOOR 72CF
Supplier: VWR International
VWR* Stainless Steel Door Storage Refrigerator^ Three swinging doors. 2038.4L. Exterior: 201Hx198Wx75Dcm. Range: 1 to 4[degree]C. 1/2 hp compressor. Anodized aluminum cabinet and interior. Three adjustable shelves per door, door lock, thermometer, and 12.7cm casters. UL/CSA/NSF. 115V.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR REFRIGERATOR SWG DOOR 72CF 89239-068 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty