Would you like to log in again?
Eppendorf 5427R Microcentrifuges
Supplier: Eppendorf
022620700 022620703 022620700 022620701 5409000136 022620702 022620702 022620703 022620701 022620704 5409000012
89239-128EA 14144.62 USD InStock
CA89239-128 89239-128 CA89239-126 CA89239-130 89239-132 89239-122 89239-130 89239-120 CA89239-124 89239-126 89239-124
Eppendorf 5427R Microcentrifuges
Centrifuges Microcentrifuges
Labs demanding high sample throughput will benefit from the 5427R, featuring a double-row 48-place rotor to increase efficiency and improve running time.
  • 48 x 1.5/2.0 mL capacity rotor available
  • Eppendorf QuickLock lids come standard with all aerosol-tight rotors
  • Automatic rotor recognition and imbalance detection for maximum safety
  • Maintains constant 4°C at maximum speed
  • FastTemp program is optimized for super-fast 11-minute pre-cooling and maximum temperature accuracy inside the rotor
The 5427R provides refrigeration with innovative compressor technology that continuously cools even after the run, so your samples stay safe.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty