Would you like to log in again?
Rotors for Centrifuges 5427 R
Supplier: Eppendorf
5409715003 5409702009 5409704001 5409714007 5409704001 5409710001 5409702009 5409708007 5409712004 5409715003 5409700006 5409714007 5409706004 5409700006 5409710001 5409706004
89239-150EA 2797.72 USD InStock
89239-150 89239-140 CA89239-142 89239-144 CA89239-150 89239-142 CA89239-134 89239-136 89239-148 89239-134 89239-146 CA89239-138 89239-138 CA89239-136 CA89239-148 CA11028-848
Rotors for Centrifuges 5427 R
Centrifuges Rotors
Eppendorf rotors for centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
Elevate your centrifugation experience with Eppendorf rotors crafted for centrifuge 5427 R. These precision rotors offer diverse capacities and speeds, providing flexibility for microcentrifugation or larger sample volumes. Accessories such as adaptors and buckets enhance customization for specific applications. Maximize efficiency with reliable sample separation, meeting the demands of molecular biology, genomics, and more. Explore the full range of options to tailor your centrifuge setup, achieving reproducible results and optimizing your laboratory workflow.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty