Would you like to log in again?
Rotors for Minispin®/Minispin® Plus Centrifuges
Supplier: Eppendorf
022665821 022668498 022665821
97058-978EA 548 USD InStock
97058-978 CA022665821 89239-368
Rotors for Minispin®/Minispin® Plus Centrifuges
Centrifuges Rotors
Eppendorf Rotors for MiniSpin/MiniSpin plus, precision for mini centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with versatile adaptors and buckets for efficient sample processing in a compact format.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
Elevate your mini centrifugation experience with Eppendorf Rotors designed for MiniSpin/MiniSpin plus. These rotors offer a range of capacities and speeds, catering to diverse sample processing needs. Accessorize your setup with versatile adaptors and buckets, enhancing the flexibility of your mini centrifuge for various applications. Achieve precision in your experiments within the compact format, ensuring reliable and efficient sample separation. Explore the comprehensive range to optimize your MiniSpin/MiniSpin plus for reproducible results.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty