Would you like to log in again?
9 mm Glass Screw-Thread Autosampler Vial Kits, Advanced Vial Closure System (AVCS), Thermo Scientific
Supplier: Thermo Scientific
C5000-194W C5000-80 C5000-94W C5000-95W C5000-82P C5000-972 C5000-LV95 C5000-80W C5000-595W C5000-195W C5000-992 C5000-592 C5000-186WR C5000-186 C5000-94P C5000-586W C5000-76W C5000-95 C5000-54P C5000-580 C5000-184W C5000-92 C5000-192 C5000-580W C5000-82W C5000-88W C5000-188W C5000-86W C5000-186W C5000-387 C5000-592W C5000-78AW C5000-183W C5000-196W C5000-180 C5000-182W C5000-93W C5000-86 C5000-91W C5000-92P C5000-92W C5000-178 C5000-180W C5000-188WR C5000-83W C5000-192W C5000-54A C5000-97
76047-572PK 60.77 USD InStock
76047-572 10222-296 89239-440 10222-310 89239-444 89239-442 10119-716 82428-560 10119-738 89239-438 10119-718 89239-436 82428-564 82428-562 10119-730 10119-732 10119-714 10119-736 10222-182 10119-690 10119-692 10222-188 10222-200 89239-452 89239-450 10222-202 89239-454 10119-726 89239-448 89239-446 10119-728 10135-750 82428-554 10119-808 76047-538 82428-558 10119-740 10119-686 10119-720 10119-742 82428-556 10119-688 10119-744 10119-722 10766-732 10119-746 10222-180 10119-724
9 mm Glass Screw-Thread Autosampler Vial Kits, Advanced Vial Closure System (AVCS), Thermo Scientific
Vials
These autosampler vial kits are available with assembled or unassembled closures.
  • Save time during sample preparation and reduce the risk of contamination
Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers.
Delivery information: Each kit includes 100 glass vials and 100 screw thread caps with pre-assembled septa.
Packaging: The kit is packaged in convenient vial trays with clear covers or in economical polybags.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty