Would you like to log in again?
VWR® RiboReady™ Color RNA Ladders
Supplier: Avantor
VWR®
VWRV1B1492-25RXN VWRV1B1491-25RXN
89239-528EA 622.54 USD InStock
89239-528 89239-530
VWR® RiboReady™ Color RNA Ladders
Electrophoresis Reagents Molecular Weight Markers
RiboReady™ Color Micro RNA Ladder is made for precise sizing of RNA molecules in denaturing acrylamide gels
RiboReady™ Color 1KB RNA Ladder is made for precise sizing of RNA molecules in denaturing agarose gels. RiboReady™ Color 1KB RNA Ladder is supplied in a ready-to-use mixture that does not require heating or addition of a denaturing agent before use. RiboReady™ Color 1KB RNA Ladder has the added advantage of containing pre-stained single-strand nucleic acids that are visible during electrophoresis. The apparent sizes of bands in the RiboReady™ Color 1KB RNA Ladder are 200, 500, 1,000, 2,000, 4,000 and 8,000 bases in length.
RiboReady™ Color Micro RNA Ladder is supplied in a ready-to-use mixture that does not require heating or addition of a denaturing agent before use. RiboReady™ Color Micro RNA Ladder has the added advantage of containing pre-stained single-strand nucleic acids that are visible during electrophoresis. The apparent sizes of bands in the RiboReady™ Color Micro RNA Ladder are 100, 75, 50, 40, 30 and 20 bases in length.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty