Would you like to log in again?
Square Cover Glasses, Corning®
Supplier: Corning
Corning®
2855-18 2855-25 2850-22 2850-18 2845-22 2855-22 2845-18 2845-25 2850-25
89239-702CS 131.26 USD InStock
89239-702 89239-700 89239-706 89239-690 89239-704 89239-694 89239-692 89239-698 89239-696
Square Cover Glasses, Corning®
Cover Glasses
These square cover glasses are designed for non sterile applications.
  • Packed one ounce per pack and 10 packs per case
Special, optically clear, water-white, borosilicate cover glass is resistant to surface attachment or weathering and will remain clear for extended periods of time.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty