Would you like to log in again?
VWR® Microprobe Set
Supplier: Avantor
11648-276
89259-848EA 126.21 USD InStock
89259-848
VWR® Microprobe Set
Dissection Needles
This VWR® Microprobe Set is for micromanipulation in the lab.
  • Stainless steel construction
  • Needle-sharp tips
  • Variety of microprobes available
Each microprobe is precision-ground stainless steel that has been tempered for quality control and experimental requirements. Tips are needle-sharp and have been hand-honed. Set includes six different microprobes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty