Would you like to log in again?
VWR® Micro Spatulas, Double-Ended
Supplier: Avantor
VWR®
11648-295 11648-297 11648-296
89259-886EA 27.22 USD InStock
89259-886 89259-890 89259-888
VWR® Micro Spatulas, Double-Ended
Spatulas
These VWR® Micro spatulas are double-ended and stainless steel.
  • Stainless steel construction
  • Double-ended design
Spatula has a curved spatula. Spatula is sharpened at the tip for cutting and separating and has a small knob at other end for crushing crystals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty