Would you like to log in again?
Dry Block Heater, Standard Well Plates, Glas-Col
Supplier: Glas-Col
099A HBC12
89260-008EA 2093.25 USD InStock
89260-008
Dry Block Heater, Standard Well Plates, Glas-Col
Block Heaters Digital Block Heaters
The Dry Block Heater is a convenient answer for those needing a small constant temperature heater.
  • User friendly digital control
  • Small constant temperature heater
  • Heater opening is standard SBS format
Equipped with a digital control with easy setup and simple input commands. Ambient temperature range 32 -149 °F. Dual 4 digit display.
Ordering information: Base wih single SBS format block.
Delivery information: 7-10 days after order.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty