Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MC1061 F-Competent Cells, Biosearch Technologies
Supplier: Biosearch Technologies
60514-1 60514-2
89260-194EA 466.59 USD InStock
89260-194 89260-196
MC1061 F-Competent Cells, Biosearch Technologies
Competent Cells
MC1061 F- Electrocompetent Cells
An amber suppressor strain, identical to SS320 except that it lacks the F′episome. Highly efficient (≥4 × 1010 cfu/µg) electrocompetent cells for general cloning or Phage Display. Note that as an F- strain, it cannot be used for re-infection by filamentous phage
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty