Would you like to log in again?
STERI-OIL® 200, Veltek Associates
Supplier: Veltek
SO200-A1Z
89260-580CS 2179.81 USD InStock
89260-580
STERI-OIL® 200, Veltek Associates
Lubricants
STERI-OIL 200 is a sterile mineral oil lubricant used for lubricating machinery in aseptic areas.The heavy consistency provides lubrication and prevents metal to metal contact, withstands high friction without displacement, and reduces metal fatigue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty