Would you like to log in again?
SP Bel-Art Switch-Grid™ Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F18745-2000 F18745-1600 F18745-3000 F18745-0045 F18745-1300 F18745-0011 F18745-0013 F18745-0012 F18745-0014 F18745-2500 F18745-0010
89260-598CS 319.73 USD InStock
89260-598 89260-610 89260-596 89260-602 89260-600 89260-612 89260-606 10755-228 89260-604 89260-594 89260-608
SP Bel-Art Switch-Grid™ Test Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
This innovative, split-frame rack holds up to two different size tubes (10–30 mm) in the same rack and allows users to quickly switch grids to hold other sizes as needed.
  • Split-frame rack design
  • Interchangeable grids
  • Color-coded for quick identification
  • Polyoxymethylene plastic rack frame
  • Chemical-resistant
  • Non-slip rubber feet
More compact and less expensive than purchasing multiple racks, their interchangeable, color-coded grids correspond to a specific test tube size ranging from 10 mm to 30 mm in diameter.
Each rack frame includes a set of grids in one size. Grid sets in alternate sizes can be purchased separately. Chemical resistant polyoxymethylene plastic rack frame and grids can be cleaned with mild detergent, ethanol, or isopropyl alcohol, and can be autoclaved at 121°C (250°F). Rubber feet keep racks from sliding on benchtop, and provide secure stacking while in storage.
Overall rack frame dimensions are 31.8L x 11.1W x 6.7H cm (12½ x 4⅜ x 2⅝").
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty