Would you like to log in again?
Qlicksmart® Blade Removal System, MYCO Medical
Supplier: MYCO Medical
Qlicksmart®
QFRGEN
89260-788CS 814.3 USD InStock
89260-788
Qlicksmart® Blade Removal System, MYCO Medical
Blade Removers and Dispensers
OSHA compliant surgical blade removal system.
  • Single handed, puncture and spill proof for easy, convenient and safe blade removal
  • Audible click helps confirm proper blade removal
  • Separate bracket allows for wall or cart mounting
Compatible with most brands of general surgery blades.
Ordering information: Qlicksmart® Bracket is sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty