Would you like to log in again?
PBS Phosphate Buffered Saline Tablets, Genetex
Supplier: GeneTex
GTX48887
89284-786EA 176.53 USD InStock
89284-786
PBS Phosphate Buffered Saline Tablets, Genetex
Buffers Electrophoresis Buffers Washing Buffers
Pre-measured Phosphate Buffered Saline Tablets
Each tablet prepares a 1X PBS solution when dissolved in 100 mL of ddH2O. For in vitro research use only. Not intended for any diagnostic or therapeutic purpose. Not suitable for human or animal consumption. Store at room temperature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty