Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FTO Goat Polyclonal Antibody

Supplier: GeneTex
GTX88551
89296-536EA 607.87 USD InStock
89296-536
Anti-FTO Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to FTO (Mouse)

Approx. 60kDa band observed in Human Pancreas lysates (calculated MW of 58.0kDa according to NP_036066.2). Recommended concentration: 0.03-0.1µg/ml.

Type: Primary
Antigen: Fto
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty