Would you like to log in again?
Rotors for Centrifuges, 5810/5810 R
Supplier: Eppendorf
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
022638807 5820710004 5820740000 5820754001 022638009 022638602 022638807 022638041 022638611 022638602 5820755008 5820740000 5820755008 022638041 5820754001 022638009
47745-376EA 6711.72 USD InStock
47745-376 CA11017-374 CA47745-376 CA11017-378 CA47743-060 47743-060 53513-872 CA11017-380 CA53513-872 89305-190 CA53513-866 89305-192 75875-114 53513-866 89305-186 97058-942
Rotors for Centrifuges, 5810/5810 R
Centrifuges Rotors
Eppendorf Rotors for centrifuges 5810/5810 R - precision redefined. Tailor your high-capacity centrifugation with varied rotor options. Choose from a range of capacities and speeds, accessorize with versatile buckets, and ensure efficient sample processing. Ideal for diverse applications, guaranteeing accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
Redesign your high-capacity centrifugation with Eppendorf rotors crafted for centrifuges 5810/5810 R. These rotors offer a spectrum of capacities and speeds, providing adaptability for various sample processing needs. Accessorize with versatile buckets, ensuring efficient customization of your centrifuge setup. Ideal for diverse applications, the Centrifuge 5810 series is further enhanced with these precision-engineered rotors, guaranteeing accuracy and reproducibility in every spin. Explore the range to optimize your centrifugation experience.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty