Would you like to log in again?
Rotors and Accessories for Eppendorf®, Rotor Adapter, Multipurpose Centrifuges 5804, 5804R, 5810 and 5810R
Supplier: Eppendorf
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
022637428 5820742003 022639909 5820751002 022638614 022638564 022638614 022638866 5820757000 5920701002 022638033 5820711000 022637436 5820711000 022638629 5825719000 022638084 5804747003 022638068 022637461 5820715006 022638661 5820761008 022637452 5820716002 022638661 5804747003 022638866 022639913 022639913 022639907 022638033 5820751002 022637100 022639911 5820747005 022638930 022638937 5920700006 022638653 022638505 022638937 5820741007 5820742003 022639901 022637428 5820756004 022637002 5820741007 5920717006 022639909 022638556 022638840 022639911 5820713003 5820749008 022638840 022639905 022637436 022662970 022638935 022638653 022639907 5820712007 5820756004 5820713003 022638657 5820712007 022637401 5820708000 022639905 022638050 022638607 5820705000 022638657 022637207 022637452 5820716002
47743-074PK 472.95 USD InStock
CA80078-568 CA11017-376 CA53574-220 CA77888-208 47743-074 47743-070 75874-922 CA53513-816 89305-188 CA80078-570 CA53513-818 75874-924 89305-268 CA11017-460 CA11017-384 CA11017-386 CA11017-464 CA53513-840 75875-110 53513-874 75875-112 CA11017-456 89305-210 89305-212 97058-964 CA5820713003 75874-870 89305-238 97058-946 97058-944 CA11017-396 CA11017-430 CA11017-398 75874-872 CA11017-432 CA89030-294 53513-871 89213-148 CA53513-871 53513-868 75875-122 75875-124 75875-168 75874-988 CA5820712007 CA11017-424 53513-904 89305-242 CA11017-426 89305-244 CA5820716002 89305-200 75874-902 97058-950 80078-568 97058-938 CA022638614 CA11017-400 CA77888-214 75875-126 89030-294 CA47743-074 CA47743-070 CA5820715006 75875-178 89305-272 75875-136 CA53513-820 CA53513-904 89305-196 CA022638657 89305-276 80078-570 CA53513-868 89305-198 89173-646 CA5820711000 75875-172
Rotors and Accessories for Eppendorf®, Rotor Adapter, Multipurpose Centrifuges 5804, 5804R, 5810 and 5810R
Centrifuges Rotors
Various adapters, rotors, and other accessories for centrifuges 5804 and/or 5804R and/or 5810 and/or 5810R. Eppendorf Rotors for Centrifuges 5804(R) and 5810(R) precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
Experience precision in high-performance centrifugation with Eppendorf Rotors designed for Centrifuge 5804/5804 R. These rotors offer diverse capacities and speeds, providing flexibility for various sample processing needs. Accessorize with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-performance applications, the Centrifuge 5804 series is further optimized with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your centrifugation experience.
Ordering information: Rotors, adapters, and accessories are only for use with centrifuge models specified. Unless otherwise specified, rotors may be used with all centrifuge models.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty