Would you like to log in again?
STEM CELL FACTOR SCF HUMAN
Supplier: Genscript
STEM CELL FACtoR SCF HUMAN^ SCF is a hematopoietic growth factor that exerts its activity during the early stages of hematopoiesis. SCF stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in Bone marrow cultures and act synergistically with colony stimulating factors
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STEM CELL FACTOR SCF HUMAN 89305-388 Z02692-10 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty