Would you like to log in again?
INTERLEUKIN-16 IL-16 HUMAN
Supplier: Genscript
INTERLEUKIN-16 IL-16 HUMAN^ IL-16 is a CD8+ T cell-derived cytokine that induces chemotaxis of CD4+ T cells and CD4+ monocytes and eosinophils. Analysis By gel filtration suggests that, under physiological conditions, human IL-16 exists predominantly asa noncovalently linked multimer
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INTERLEUKIN-16 IL-16 HUMAN 89305-560 Z02196-50 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty