Would you like to log in again?
Human VEGF165 (from P. pastoris)
Supplier: GenScript Biotech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Z02689-10 Z02689-100 Z02689-1 Z02689-50
89305-590EA 423.61 USD InStock
89305-590 89305-592 89305-594 89305-588
Human VEGF165 (from P. pastoris)
Proteins and Peptides
Vascular endothelial growth factor (VEGF) is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, Vascular endothelial growth factor (VEGF) plays a prominent role in normal and pathological angiogenesis.
Substantial evidence implicates VEGF in the induction of tumor metastasis and intra-ocular neovascular syndromes. VEGF signals through the three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant human Vascular Endothelial Growth Factor A165 (rhVEGF-A165) produced in Pichia pastoris is a disulfide-linked homodimer containing two polypeptide chains of 165 amino acids each. A fully biologically active molecule, rhVEGF-A165 has a molecular mass of 38.2kDa.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty