Would you like to log in again?
INTERLEUKIN-13 VARIANT IL-13
Supplier: Genscript
INTERLEUKIN-13 VARIANT IL-13^ The human IL-13 cDNA encodes a 132 amino acid protein containing a proposed 20 amino acid signal peptide. Human IL-13 shares approximately 30% amino acid sequence homology to human IL-4 and the two cytokines exhiBit overlapping Biological activities
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INTERLEUKIN-13 VARIANT IL-13 89305-646 Z02711-10 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty