Would you like to log in again?
THYMIC STROMAL LYMPHOPOIETIN
Supplier: Genscript
THYMIC STROMAL LYMPHOPOIETIN^ TSLP is a hemopoietic protein that is expressed in the heart, liver and prostate. TSLP overlaps Biological activities with IL-7 and Binds with the heterodimeric receptor complex consisting of the IL-7R alpha chain (IL-7R? and the TSLP-specific chain (TSLPR).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
THYMIC STROMAL LYMPHOPOIETIN 89305-688 Z02721-10 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty