Would you like to log in again?
SOLUBLE CD40 LIGAND SCD40L HU
Supplier: Genscript
SOLUBLE CD40 LIGAND SCD40L HU^ CD40 ligand, CD40L (also known as CD154, TRAP or gp39), is a 261 amino acid type II transmemBrane glycoprotein Belonging to the TNF family, CD40L is expressed predominantly on activated CD4+ T lymphocytes
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SOLUBLE CD40 LIGAND SCD40L HU 89305-712 Z02727-50 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty