Would you like to log in again?
B-CELL MATURATION ANTIGEN BCM
Supplier: Genscript
B-CELL MATURATION ANTIGEN BCM^ BCMA, a memBer of the TNF receptor superfamily, Binds to BAFF and APRIL. BCMA is expressed on mature B-cells and other B-cell lines and plays an important role in B cell development, function and regulation. BCMA also has the capaBility to activate NF-kappaB and JNK
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
B-CELL MATURATION ANTIGEN BCM 89305-726 Z02731-1 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty