Would you like to log in again?
BETA-DEFENSIN 2 BD-2 HUMAN
Supplier: Genscript
BETA-DEFENSIN 2 BD-2 HUMAN^ Defensins (alpha and Beta) are cationic peptides with a Broad spectrum of antimicroBial activity that comprise an important arm of the innate immune system. The A-defensins are distinguished from the B-defensins By the pairing of their three disulfide Bonds.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BETA-DEFENSIN 2 BD-2 HUMAN 89305-812 Z02755-20 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty