Would you like to log in again?
INTERLEUKIN-33 IL-33 MOUSE
Supplier: Genscript
INTERLEUKIN-33 IL-33 MOUSE^ IL-33 is a proinflammatory protein that shares structural and functional characteristics with the IL-1 cytokine family. It Binds and signals through the IL-1RL1/ST2 receptor activating NF-kappaB and MAP kinases. IL-33 inducesproduction of TH2 cell related cytokines
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INTERLEUKIN-33 IL-33 MOUSE 89305-872 Z02772-1 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty