Would you like to log in again?
SDF-1 ALPHA/CXCL12 HUMAN
Supplier: Genscript
SDF-1 ALPHA/CXCL12 HUMAN^ SDF-1A and SDF-1B memBers of the chemokine alpha suBfamily that lack the ELR domain, were identified using the signal sequence trap cloning strategy from a mouse Bone-marrow stromal cell line. The cDNAs encode precursor proteins of 89 and 93 amino acid residues
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SDF-1 ALPHA/CXCL12 HUMAN 89306-004 Z02825-1 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty