Would you like to log in again?
RANTES/ CCL5 HUMAN
Supplier: Genscript
RANTES/ CCL5 HUMAN^ CCL5 or RANTES was initially discovered By suBtractive hyBridization as a transcript expressed in T cells But not B cells. Eosinophilchemotactic activities released By thromBinstimulated human platelets have also Been purified and found to Be identical to RANTES
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RANTES/ CCL5 HUMAN 89306-030 Z02832-20 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty